Cationic starch N 0,4%
Supplier: SPECTRUM CHEMICAL MFG CORP
S1191-250MG
S1191-100MG
S1191-1GM
700015-664EA
529
USD
700015-664
700015-666
700015-662
Cationic starch N 0,4%
Cationic starch
Cationic Starch, Nitrogen 0.4 Percent is a carbohydrate and polysaccharide that has a large number of glucose units that are linked by glycosidic bonds that has been modified by reaction with a quaternary ammonium salt. These products are used to increase the dry strength and emulsification of sizing agents.
Storage Temperature:
Ambient |
CAS Number:
56780-58-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...