Carbamazepine ≥98.0% USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 5H-Dibenzo[b,f]azepine-5-carboxamide

CA250-5GM CA250-25GM CA250-100GM CA250-1KGBL
95032-374EA 3445 USD
95032-374 95032-372 95032-378 95032-376
Carbamazepine ≥98.0% USP
Carbamazepine

Carbamazepine, USP is an anti-seizure and mood-stabilizing drug. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Exclamation mark;Health hazard

Formula: C₁₅H₁₂N₂O
MW: 236.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005073
CAS Number: 298-46-4

Order Now

Specification Test Results

Assay (Dried Basis) 98.0 - 102.0%
X-Ray Diffraction Conforms
Acidity To Pass Test
Alkalinity To Pass Test
Loss On Drying max. 0.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR