Calcium undecylenate ≥98.0%

C1540-25KGBL C1540-2.5KG C1540-500GM
700013-778EA 1123.76 USD
700013-778 700013-780 700013-782
Calcium undecylenate ≥98.0%
Calcium undecylenate

Calcium Undecylenate is a chemical intermediate; preservative, bacteriostat, and fungistat.


Non-graded Calcium Undecylenate supplied by Spectrum is indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Storage Temperature: Ambient
MDL Number: MFCD00068309
CAS Number: 1322-14-1

Order Now

Specification Test Results

Assay (Dried Basis) 98.0-102.0 %
Loss On Drying 2.0 - 5.7 %
Free Undecylenic Acid max. 0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR