Calcium saccharin ≥98.0% FCC

C1146-12KG C1146-500GM C1146-2.5KG
95035-582EA 1683.78 USD
95035-582 75810-774 95035-584
Calcium saccharin ≥98.0% FCC
Calcium saccharin

Calcium Saccharin, FCC is an artificial sweetener that is about 300 times as sweet as table sugar (sucrose). Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Formula: C₁₄H₈CaN₂O₆S₂·3½H₂O
Storage Temperature: Ambient
CAS Number: 6381-91-5

Order Now

Specification Test Results

Assay (Anhydrous Basis) 98.0 - 101.0 %
Benzoate and Salicylate To Pass Test
Lead max. 2 mg/kg
Readily Carbonizable Substances To Pass Test
Selenium max. 0.003 %
Toluenesulfonamides To Pass Test
Water Max. 15%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR