Calcium glycerophosphate ≥98.0% FCC
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Calcium 2,3-dihydroxypropyl phosphate
, Glycerophosphoric Acid Calcium Salt
CA147-2.5KG
CA147-12KG
CA147-125GM
CA147-50KGBL
CA147-500GM
95032-282EA
467
USD
95032-282
95032-280
700013-950
700013-952
95032-278
Calcium glycerophosphate ≥98.0% FCC
Calcium glycerophosphate
Calcium Glycerophosphate, FCC is used to neutralize acidic foods when consumed. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
C₃H₇CaO₆P MW: 210.13 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00042643 CAS Number: 27214-00-2 |
Specification Test Results
Assay (C₃H₇CaO₆P) | 98.0 - 100.5 % |
Alkalinity | To Pass Test |
Loss On Drying | max. 12.0 % |
Lead (Pb) | max. 4 mg/kg |
Identification | To Pass Test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...