Butamben (butyl-4-aminobenzoate) ≥98.0% USP
Synonyms:
Butyl 4-aminobenzoate
B1012-50KGBL
700007-234EA
62947.41
USD
700007-234
Butamben (butyl-4-aminobenzoate) ≥98.0% USP
Butamben (Butyl 4-aminobenzoate)
All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specification Test Results
Assay (Dried Basis) | 98.0 - 101.0 % |
Completeness and Color of Solution | To Pass Test |
Melting Range | 57 - 59 °C |
Reaction | To Pass Test |
Loss On Drying | max. 1.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...