Budesonide ≥98.0%, micronized Ph. Eur.

Synonyms: (+)-16alpha,17alpha-Butylidenedioxy-11beta,21-dihydroxy-1,4-pregnadiene-3,20-dione

B1595-1KGBL B1595-1GM B1595-500MG B1595-5GM
95032-162EA 30214.8 USD
95032-162 95032-160 95032-166 95032-164
Budesonide ≥98.0%, micronized Ph. Eur.
Budesonide

Budesonide, Micronized, EP is a glucocorticoid steroid anti-inflammatory used to treat certain bowel conditions. Spectrum Chemical EP products, similar to USP products, are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities


GHS hazard : Exclamation mark;Health hazard

Formula: C₂₅H₃₄O₆
MW: 430.54 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00083259
CAS Number: 51333-22-3

Order Now

Specification Test Results

Assay (Dried Basis) 98.0 - 102.0%
Related Substances To Pass Test
Epimer A To Pass Test
Methanol (Ch₃OH) max. 0.1%
Loss On Drying max. 0.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR