Buchu leaf oil

B1655-5ML
700013-632EA 116.22 USD
700013-632
Buchu leaf oil
Buchu leaf oil

Buchu Leaf Oil

Storage Temperature: Ambient
MDL Number: MFCD00676571
CAS Number: 68650-46-4

Order Now

Specification Test Results

Appearance Yellow to Browni

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR