Brilliant black BN

Synonyms: Black PNBrilliant Black ACI 28440Blue Black BNFood Black 1

B3115-25GM B3115-10GM
700010-242EA 198.53 USD
700010-242 700010-240
Brilliant black BN
Brilliant black BN

Brilliant Black BN is a non-food colorant also known as Brilliant Black BN.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₂₈H₁₇N₅O₁₄S₄
MW: 867.66 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00078980
CAS Number: 2519-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR