β-Ionone (synthetic) ≥96%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: βeta-Jonone4-(2,6,6-Trimethylcyclohex-1-en-1-yl)but-3-en-2-one

I2106-100GM I2106-25GM
700014-580EA 142.18 USD
700014-580 700014-578
β-Ionone (synthetic) ≥96%
β-Ionone

beta-Ionone, Synthetic is used as to make Vitamins A, E and K1 and as a fragrance in perfumes, cosmetics and personal care products, and household cleaners and detergents.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₃H₂₀O
MW: 192.30 g/mol
Density: 0.945
Storage Temperature: Ambient
CAS Number: 14901-07-6

Order Now

Specification Test Results

Assay Approx. 96 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR