Berberine sulfate hydrate

Supplier: SPECTRUM CHEMICAL MFG CORP

B2755-50GM
700009-604EA 572 USD
700009-604
Berberine sulfate hydrate
Berberine sulfate hydrate

Berberine Sulfate has various beneficial effects on the cardiovascular system and significant anti-inflammatory activities.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₄₀H₃₆N₂O₁₂S
MW: 768.79 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00031743
CAS Number: 316-41-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR