Benzoyl peroxide 75%, hydrous ≥75% USP
Supplier: SPECTRUM CHEMICAL MFG CORP
|
|
Synonyms:
Dibenzoyl peroxide
BE156-12KGBL
BE156-500GM
BE156-25GM
BE156-45KGBL
BE156-1KGBL
BE156-125GM
BE156-10KGBL
75810-704EA
717
USD
75810-704
75810-702
75810-700
95030-902
95030-900
95030-898
75810-706
Benzoyl peroxide 75%, hydrous ≥75% USP
Benzoyl peroxide
Hydrous Benzoyl Peroxide, 75 Percent, USP is used to treat mild to moderate acne. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities
GHS hazard : Flame;Flame over circle;Exclamation mark
UN3104
Formula:
C₁₄H₁₀O₄ Storage Temperature: Ambient |
MDL Number:
MFCD00003071 CAS Number: 94-36-0 UN: 3104 ADR: 5.2,II |
Specification Test Results
Assay (Benzoyl Peroxide) | 70.0 - 80.0 % |
Organic Impurities | To Pass Test |
Identification | To Pass Test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...