Benzocaine (ethyl-4-aminobenzoate) ≥98.0% USP
Supplier: SPECTRUM CHEMICAL MFG CORP
|
|
Synonyms:
Aethoform
, Ethyl 4-aminobenzoate
BE130-12KG
BE130-5GM
BE130-2.5KG
BE130-125GM
BE130-25GM
BE130-1GM
BE130-500GM
95031-980EA
444
USD
95031-980
89078-604
95031-976
75810-696
95031-978
75810-694
75810-692
Benzocaine (ethyl-4-aminobenzoate) ≥98.0% USP
Benzocaine (Ethyl 4-aminobenzoate)
Benzocaine, USP is a mild local anesthetic. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities
GHS hazard : Exclamation mark
Formula:
C₉H₁₁NO₂ MW: 165.19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00007892 CAS Number: 94-09-7 |
Specification Test Results
Assay (Dried Basis) | 98.0 - 102.0 % |
Loss On Drying | max.1.0 % |
Residue On Ignition | max. 0.1 % |
Chloride | To Pass Test |
Heavy Metals | max. 10 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...