Bay oil FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

B1061-100ML B1061-25ML B1061-500MLGL
75810-658EA 176.27 USD
75810-658 75810-660 95030-742
Bay oil FCC
Bay oil

Bay Oil, FCC is used in cooking as a flavoring agent. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Flame;Exclamation mark
UN1197

Storage Temperature: Ambient
MDL Number: MFCD00130604
CAS Number: 8006-78-8
UN: 1197
ADR: 3,III

Order Now

Specification Test Results

Assay (As Phenols; By Volume) 50 - 65 %
Angular Rotation max. -3
Refractive Index @ 20°C 1.507- 1.516
Specific Gravity @ 25°C 0.950- 0.990
Identification To Pass Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR