Basil oil European Type
Supplier: SPECTRUM CHEMICAL MFG CORP
|
|
B1311-25ML
B1311-500ML
700010-454EA
1412
USD
700010-454
700010-452
Basil oil European Type
Basil oil
Basil Oil, European Type is aromatic, spicy perfuming ingredient with a licorice (anise) scent.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Storage Temperature:
Ambient |
MDL Number:
MFCD00217166 CAS Number: 8015-73-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...