Azathioprine ≥98.0% USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 6-(1-Methyl-4-nitroimidazol-5-yl)thiopurine

A1383-100GM A1383-25GM A1383-5GM A1383-1KGBL
75810-538EA 3711 USD
75810-538 75810-546 75810-544 75810-542
Azathioprine ≥98.0% USP
Azathioprine

Azathioprine, USP is an immunosuppressive drug used in autoimmune diseases and organ transplants. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Exclamation mark;Health hazard

Formula: C₉H₇N₇O₂S
MW: 277.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00069203
CAS Number: 446-86-6

Order Now

Specification Test Results

Assay (Dried Basis) 98.0 - 102.0 %
Loss On Drying max.1.0 %
Residue On Ignition max. 0.1 %
Organic Impurities To Pass Test
Identification To Pass Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR