Asiaticoside ≥90%

Supplier: SPECTRUM CHEMICAL MFG CORP

A3649-100MG
700009-484EA 1183 USD
700009-484
Asiaticoside ≥90%
Asiaticoside

Asiaticoside is used to study potential treatments for wounds and burns. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Storage Temperature: Ambient
MDL Number: MFCD06642601
CAS Number: 16830-15-2

Order Now

Specification Test Results

Assay (HPLC) 90% min.
Loss On Drying max. 5%
Heavy Metals max. 20 ppm
Sulfated Ash max. 0.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR