SPCMAM162
AM162-25GM
75810-630EA 62.77 USD
75810-630
SPCMAM162
AMINOPHYLLINE, ANHYDROUS, POWDER, USP is usually found as a dihydrate and is most commonly used for treating airway obstructions due to COPD or asthma. The USP grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
GHS hazard : Skull and crossbones
UN2811