Aluminum chlorohydrate dihydrate USP
Aluminum Chlorohydrate, Dihydrate, USP is a group of specific aluminum salts used in topical skin products (deodorants and antiperspirants) as an astringent in the management of hyperhidrosis. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities
It appears as a colorless to yellow or white, odorless, solid crystalline powder. The USP grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
Al₂ClH₅O₅·2H₂O Storage Temperature: Ambient |
MDL Number:
MFCD00053388 CAS Number: 12359-72-7 |
Specification Test Results
Anhydrous Aluminum Chlorohydrate | As Reported |
Content of Aluminum | As Reported |
Content of Chloride | As Reported |
Aluminum/Chloride Atomic Ratio | 1.91:1- 2.10:1 |
pH of a 15 in 100 (w/w) Solution | 3.0 - 5.0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...