D-ɑ-Tocopherylquinone

Synonyms: Vitamin E Quinone (D-form)

T2546-1GM
700016-032EA 1775.43 USD
700016-032
D-ɑ-Tocopherylquinone
D-alpha-Tocopherylquinone

alpha-Tocopherylquinone is a vitaman E oxidation product with antioxidant activities.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₂₉H₅₀O₃
MW: 446.72 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00135657
CAS Number: 7559-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR