α-Methylstyrene vinyl toluene copolymer

Supplier: SPECTRUM CHEMICAL MFG CORP

M1654-250GM M1654-1KG M1654-50GM
700012-256EA 131.78 USD
700012-256 700012-254 700012-252
α-Methylstyrene vinyl toluene copolymer
alpha-Methylstyrene vinyl toluene copolymer

alpha-Methylstyrene-vinyl Toluene Copolymer is used as an adhesive, paint and lacquer additive for viscosity control. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Storage Temperature: Ambient
MDL Number: MFCD00284312
CAS Number: 9017-27-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR