Aloin, powder

Synonyms: (S)-1,8-Dihydroxy-3-(hydroxymethyl)-10-((2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)anthracen-9(10H)-one10-β-D-Glucopyranosyl-1,8-dihydroxy-3-(hydroxymethyl)-9(10H)-anthracenone1,8-Dihydroxy-10-(beta-D-glucopyranosyl)-3-(hydroxymethyl)-9(10H)-anthracenoneAloin ABarbaloin

AL217-50KGBL
700013-552EA 10507.62 USD
700013-552
Aloin, powder
Aloin

Aloin, Powder, also known as Barbaloin, can be used as a stimulant-laxative.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₂₁H₂₂O₉
MW: 418.398 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00151160
CAS Number: 8015-61-0

Order Now

Specification Test Results

Barbaloin Content 50 %
Loss On Drying max. 5.0 %
Escherichia Coli Negative
Salmonella Negative

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR