Allantoin ≥98%, Powder

Supplier: SPECTRUM CHEMICAL MFG CORP

AL213-2.5KG AL213-12KG AL213-50KGBL AL213-500GM AL213-100GM
700008-580EA 3770.3 USD
700008-580 700008-574 700008-572 700008-578 700008-576
Allantoin ≥98%, Powder
Allantoin

Allantoin, Powder, also referred to as glyoxyldiureide, is an important metabolic intermediate in many mammals, plants and bacteria, exept humans and higher apes. It is present in botanical extracts and in urine from most animals. Synthesized Allantoin is nature-identical, safe and non-toxic and meets CTFA and JSCI requirements. It is used in toothpaste, mouthwash and other oral hygiene products

Formula: C₄H₆N₄O₃
Storage Temperature: Ambient
MDL Number: MFCD00005260
CAS Number: 97-59-6

Order Now

Specification Test Results

Assay 98 % min.
Ash max.0.2 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR