Allantoin ≥98.5% USP
Supplier: SPECTRUM CHEMICAL MFG CORP
A2721-50KGBL
700008-570EA
6337.9
USD
700008-570
Allantoin ≥98.5% USP
Allantoin
Allantoin, USP is a moisturizing ingredient.
All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
C₄H₆N₄O₃ Storage Temperature: Ambient |
MDL Number:
MFCD00005260 CAS Number: 97-59-6 |
Specification Test Results
Assay | 98.5 - 101.0 % |
Acidity or Alkalinity | To Pass Test |
Loss On Drying | max. 0.1 % |
Organic Impurities | max. 0.5 % |
Residue On Ignition (Sulphated Ash) | max. 0.1 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...