Adenosine-5'-monophosphoric acid (AMP) free acid ≥99%

Synonyms: ((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl dihydrogen phosphate5'-AMP5'-Adenylic acidAdenosine 5'-monophosphate

AD113-100GM AD113-5KGBL AD113-25GM
700009-064EA 8217.03 USD
700009-064 700009-058 700009-060
Adenosine-5'-monophosphoric acid (AMP) free acid ≥99%
Adenosine-5'-monophosphoric acid (AMP)

ADENOSINE-5-MONOPHOSPHATE, FREE ACID, also known as Adenosine monophosphate, is used as a monomer in DNA and RNA. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Storage temperature - REFRIGERATE: Keep tightly closed.

Formula: C₁₀H₁₄N₅O₇P
MW: 347.22 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00005750
CAS Number: 61-19-8

Order Now

Specification Test Results

Assay 99 % min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR