Acesulfame potassium ≥99%
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Potassium 6-methyl-4-oxo-4H-1,2,3-oxathiazin-3-ide 2,2-dioxide
, Acesulfame K
, Acesulphame K
, Potassium 6-Methyl-1,2,3-oxathiazin-4(3H)-one 2,2-Dioxide
, 6-Methyl-1,2,3-oxathiazin-4(3H)-one 2,2-Dioxide Potassium Salt
Acesulfame Potassium is a sugar substitute that has no calories and is a white crystalline powder. Thanks to the fact that it is 200 times sweeter than sugar and stable under heat, it is used commercially as a sweetener in baking and products that need to have a long shelf life. It is also used in combination with other sweeteners in carbonated drinks and is typically used as the main sweetening agent in pharmaceutical products
It is 200 times sweeter than sugar and stable under heat, it is used commercially as a sweetener in baking and products that need to have a long shelf life. It is also used in combination with other sweeteners in carbonated drinks and is typically used as the main sweetening agent in pharmaceutical products.
Formula:
C₄H₄KNO₄S MW: 201.24 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00043833 CAS Number: 55589-62-3 |
Specification Test Results
Assay | 99 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...