8-Anilino-1-naphthalenesulfonic acid ≥95%
Supplier: SPECTRUM CHEMICAL MFG CORP
|
|
Synonyms:
8-(Phenylamino)naphthalene-1-sulphonic acid
, 8-Anilinonaphthalene-1-sulfonic acid
, 8-Anilinonaphthalene-1-sulphonic acid
, 8-(Phenylamino)naphthalene-1-sulfonic acid
8-Anilino-1-naphthalenesulfonic Acid is also known as ANS contains an amine group and a sulfonic acid. 8-Anilino-1-naphthalenesulfonic acid is a fluorescent dye with high affinity towards hydrophobic surfaces of proteins. It is useful for determining the affinity of hydrophobic ligands to their corresponding binding proteins and is used as a probe to monitor protein-folding.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₁₆H₁₃NO₃S MW: 299.35 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00003998 CAS Number: 82-76-8 |
Specification Test Results
Assay | 95 % min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...