5-Methyl-2-phenyl-2-hexenal ≥96%
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
5-Methyl-2-phenylhex-2-enal
5-Methyl-2-phenyl-2-hexenal has a cocoa aroma. It is often used in chocolate and cocoα-type flavors. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₁₃H₁₆O MW: 188.27 g/mol Density: 0.973 Storage Temperature: Ambient |
MDL Number:
MFCD00036615 CAS Number: 21834-92-4 |
Specification Test Results
Assay | 96% min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...