4-Methoxyphenylboronic acid

Synonyms: (4-Methoxyphenyl)metaboric acidDihydroxy(4-methoxyphenyl)borane4-Methoxybenzeneboronic acid

M1001-5GM M1001-25GM
10860-464EA 428.51 USD
10860-464 10860-466
4-Methoxyphenylboronic acid
4-Methoxyphenylboronic acid

GHS hazard : Exclamation mark

Formula: C₇H₉BO₃
MW: 151.96 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00039139
CAS Number: 5720-07-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR