2,4,7,9-Tetramethyldec-5-yne-4,7-diol ((±)- and meso mixture) ≥98%
Supplier: SPECTRUM CHEMICAL MFG CORP
|
|
Synonyms:
2,4,7,9-Tetramethyl-5-decyne-4,7-diol
T1290-100GM
T1290-25GM
T1290-500GM
700003-020EA
303
USD
700003-020
700003-016
700003-018
2,4,7,9-Tetramethyldec-5-yne-4,7-diol ((±)- and meso mixture) ≥98%
2,4,7,9-Tetramethyldec-5-yne-4,7-diol ((±)- and meso-mixture)
2,4,7,9-Tetramethyl-5-decyne-4,7-diol is a wetting and foam suppression agent used in cosmetics and general purpose applications.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₁₄H₂₆O₂ MW: 226.3550 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00008942 CAS Number: 126-86-3 |
Specification Test Results
Assay | Approx 98 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...