Glycyrrhetic acid ≥98%

Synonyms: 18 beta-Glycyrrhetic acid

G2170-25GM G2170-5GM
700006-636EA 118.61 USD
700006-636 700006-634
Glycyrrhetic acid ≥98%
Glycyrrhetic acid

18beta-Glycyrrhetinic Acid, also known as Enoxolone, is often used to treat peptic ulcers and also has expectorant properties.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₃₀H₄₆O₄
MW: 470.69 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003706
CAS Number: 471-53-4

Order Now

Specification Test Results

Assay Approx. 98 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR