1-(o-Tolyl)biguanide

T1314-100GM T1314-500GM
700007-948EA 230.62 USD
700007-948 700007-946
1-(o-Tolyl)biguanide
1-(o-Tolyl)biguanide

1-(o-Tolyl)-biguanide is used as a cosmetic antioxidant and in general cleaning and washing products.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₉H₁₃N₅
MW: 191.2330 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00019731
CAS Number: 93-69-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR