(±)-3-Hydroxybutyric acid

Synonyms: (±)-3-Hydroxybutanoic acid3-Hydroxybutyric acidDL-3-Hydroxybutyric Acid

H2744-5GM
10860-516EA 194.57 USD
10860-516
(±)-3-Hydroxybutyric acid
(±)-3-Hydroxybutyric acid

(+/-)-3-Hydroxybutyric Acid, or beta-Hydroxybutyric acid, is the precursor to polyesters, which are biodegradable plastics.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₄H₈O₃
MW: 104.11 g/mol
Density: 1.16
Storage Temperature: Ambient
MDL Number: MFCD00004546
CAS Number: 300-85-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR