(+)-Biotin-4-nitrophenyl ester

Synonyms: 4-Nitrophenyl 5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoate

B2570-1GM B2570-100MG B2570-250MG
10860-502EA 986.43 USD
10860-502 10860-504 10860-500
(+)-Biotin-4-nitrophenyl ester
(+)-Biotin-4-nitrophenyl ester

(+)-Biotin 4-Nitrophenyl Ester is a reagent used to synthesize biotin-labeled oligonucleotides. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₆H₁₉N₃O₅S
MW: 365.40 g/mol
Storage Temperature: Low Temperature Freezer (–40°C)
MDL Number: MFCD15141972
CAS Number: 33755-53-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR