L(-)-Bornyl acetate

Supplier: SPECTRUM CHEMICAL MFG CORP

B3414-100GM
10860-482EA 217 USD
10860-482
L(-)-Bornyl acetate
L(-)-Bornyl acetate

(-)-Bornyl Acetate is a liquid without color with a camphor-like scent. It is mainly used as an odor in the production of perfumes and in flavoring, as well as manufacturing as a plasticizer. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available

Formula: C₁₂H₂₀O₂
Density: 0,982
Storage Temperature: Ambient
MDL Number: MFCD00867808
CAS Number: 5655-61-8

Order Now

Specification Test Results

Assay Approx. 95%
Specific Rotation @ 20°C -38° to -44°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR