    Sort by:
Poloxamer 331
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Spectrum Chemicals
Description:   POLOXAMER 331 is a nonionic triblock copolymer. It is made up of a main hydrophobic chain of polyoxypropylene bordered on each side by two hydrophili...
Small Business Enterprise