    Sort by:
Lipase
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Spectrum Chemicals
Description:   LIPASE is an enzyme that is used to catalyze the hydrolysis of lipids. It is considered to be a subclass of the esterases and plays a crucial role in...
Small Business Enterprise