    Sort by:
Croscarmellose sodium
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Spectrum Chemicals
Description:   CROSCARMELLOSE SODIUM, NF is an internally crosslinked sodium carboxymethylcellulose. It is used as a super-disintegrant in pharmaceutical formulatio...
Small Business Enterprise