You Searched For: Vegetable oil brominated
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Supplier: Spectrum Chemicals
Description: Brominated Vegetable Oil, FCC, also known as BVO, is a mix of plant-derived triglycerides and is used to help emulsify citrus flavored drinks. Spectru...