    Sort by:
Aloe vera
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Spectrum Chemicals
Description:   ALOE VERA, 200:1, POWDER - Spectrum’s freeze dried ALOE VERA powder is easy to use. Just add 199 parts distilled or deionized water to 1 part aloe ve...
Small Business Enterprise