Masterflex® C/L Tygon® E-LFL, Pump Tubing, Cole Parmer
Supplier: Cole Parmer
6449-42 6449-18 6449-48 6449-30 6449-26 6449-34
10028-582PK 268.72 USD
10028-582 10028-584 10028-586 10028-588 10028-578 10028-580
Masterflex® C/L Tygon® E-LFL, Pump Tubing, Cole Parmer
Tubing Peristaltic Pump Tubing
Masterflex® Tygon® E-LFL Microbore/Autoanalysis tubing is nontoxic and transparent.
  • Autoclave or ethylene oxide
  • Good chemical resistant and oxidizing
  • Temperature range: –50 to 74 °C
  • Applications: General laboratory uses
Bends to small radius curves.
Certifications: USP Class VI requirements, EP 3.2.6, and FDA compliant.