Screw-Thread Vials, 9 mm, SUN SRi
Supplier: SUN SRi
501 500
89209-774PK 67.92 USD
89209-774
Screw-Thread Vials, 9 mm, SUN SRi
Vials
These Screw thread vials have a standard 12×32 mm profile compatible with most autosamplers.
Polyproplyene vials provide improved recovery of polar and labile compounds in many HPLC applications.