REVOLVE 3 Upright, Inverted, Brightfield Microscope, Echo Laboratories
Supplier: ECHO
JCND2001 BJSD2000
76299-572EA 20483.64 USD
76299-572 76299-598
REVOLVE 3 Upright, Inverted, Brightfield Microscope, Echo Laboratories
Revolve is the world’s first and only HYBRID microscope
  • (3) Modes included: Upright + Inverted + Brightfield
  • 12MP CMOS Brightfield camera included
  • Objectives and condensers by Olympus
  • Ultra high resolution 13” retina touch display
  • Capabilities of 2 instruments for the price of 1
Revolve is significantly easier to use. No longer are years of training and experience required to drive your microscope. You’ll feel and appreciate Revolve’s familiar app interface with responsive touch control.
This easily converts between UPRIGHT and INVERTED configurations. Eyepieces have been replaced with an iPad Pro to view and capture stunning crystal-clear images.
Accessories information: Package includes Microscope Body, iPad Pro with Locking Cradle, 12MP Brightfield Color Camera, Power Supply, Precision XY Stage, Microscope Cover, Dark Box, Stage Clip, Lens Cloth.
Objectives, Condensers, Stage Inserts are available separately.
Ordering information: Microscope includes a one-year manufacturer's warranty.
Contact your VWR Sales Representative to demo the newest innovations in microscopy.