VWR® Computer Data Acquisition System with USB Cable
Supplier: VWR, part of Avantor
8045
89140-204EA 334.25 USD
89140-204
VWR® Computer Data Acquisition System with USB Cable
Computer data acquisition system with USB cable is powerful and easy to use computer data capture program which works with Traceable® instruments with computer output.
  • Records interval readings from 1 to 10000 seconds
  • Displays minimum and maximum readings
  • 24/7 audible/visual alarm reminds user to check on data
  • It is designed to work with Windows® 98/Me/NT/2000/XP/Vista/Windows 7/Windows 8/Windows 10
Easily keeps track of results for thermometers, hygrometers, barometers, light meters, anemometers, manometers, dissolved oxygen, conductivity and more.
Readings are saved to a file that can be viewed, printed as-is, or merged in a report or spreadsheet format. Data continues to be collected in background even when user works on other software programs. Program automatically installs in less than two minutes, connect the cable to the instrument and to a USB port (or serial port) and data is captured. Multiple accessory extension cables expand cable length to 300'. Operation is straight forward plug and play.
Also allows up to eight unique Traceable® meters to simultaneously capture data and send it to directly to email to any smartphone or computer.
Ordering information: Includes a CD, a 6' cable (supplied USB, serial, and instrument connections) cable plugs into the instrument and computer.