Ward's® Essentials Paper Chromatography Lab Kit
Supplier: ALDON CORP SE
Ward's®
AD-WE2503-KIT
470225-520EA 48.49 USD
470225-520
Ward's® Essentials Paper Chromatography Lab Kit
Educational Classroom Kits and Activities
Students will perform chromatography on three different dyes in this hands-on lab.
  • Hands on lab activity
  • Kit aligned to Next Generation Science Standards MS-PS1-2
  • Instructions and materials included for 15 groups
  • Middle school
Through this lab, students will perform chromatography as a method for separating and analyzing a mixture of molecules. Chromatography will show how the molecules in a mixture travel at different speeds and separate based on several factors.