Ward's® Essentials How Much DNA? Kit
Supplier: Avantor
Ward's®
470227-324KT 68.19 USD
470227-324
Ward's® Essentials How Much DNA? Kit
Educational Classroom Kits and Activities
Examine the physical properties of DNA.
  • Hands on lab activity
  • Kit Aligned to Next Generation Science Standards MS-LS1-1
  • Instructions and materials included for 15 groups
  • Middle school
In this activity, students will extract DNA from different fruits and vegetables, using a salt/detergent solution and alcohol. Students will then compare the various yields of DNA from these different fruits and vegetables.
Ordering information: Coupon is included for redemption of perishable materials. Redeem by mail, phone, fax, email or online.
Caution: WARNING: CHOKING HAZARD–Product contains small parts, small balls, or marbles. Not for children under 3 yrs.