Microbial Preservation Plates, Watson Bio Lab
Supplier: Watson Bio Lab
176-251S 176-351S 176-551S 176-231S 176-531S 176-331S
75982-794BX 108.02 USD
75982-794 75983-022 75982-798 75983-014 75983-016 75982-796
Microbial Preservation Plates, Watson Bio Lab
Microplates Deep Well Microplates
Researchers can dry-preserve microbial stains such as yeast and E.coli on a paper chip coated with a preservation agent.
  • Deep Freeze or Dry Ice Not Required for Transportation
  • Space-Saving Plates
  • Sample Recovery Directly on PCR Plate
  • Low Contamination Risk
  • No Reagents or Salts
Samples can be sealed individually with an accessory protection seal to prevent contamination and inactivation. This enables compact and efficient preservation at normal room temperature, so it's no longer necessary to use deep freeze or dry ice for transportation.