LeukoCatch II Filter, Watson Bio Lab
Supplier: Watson Bio Lab
1883-110LC
75982-800PK 238.92 USD
75982-800
LeukoCatch II Filter, Watson Bio Lab
Filters Centrifugal Filters
LeukoCatch II removes leukocytes just by filtering, so no centrifuge is necessary.
  • Maximum Volume: 10 ml
  • Easy-to-use column type
  • LeukoCatch traps leukocytes from whole blood
  • Filtering time: Approximately 10 minutes
  • No centrifuge necessary
The LeukoCatch filter traps leukocytes specifically from whole blood. Filtering starts with natural dropping force and takes approximately 10 minutes.