Soybean oil USP
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Sojae Oleum
, Glycine Soja (Soybean) Oil
SO255-500MLG
SO255-20LT
SO255-500MLG
SO255-4LTGL
SO255-200LTB
SO255-20LT
95036-054EA
164
USD
95036-054
700000-106
CA700000-108
700000-108
95036-052
700000-110
Soybean oil USP
Soybean oil
All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Density:
0.917 Storage Temperature: Ambient |
MDL Number:
MFCD00132356 CAS Number: 8001-22-7 |
Specification Test Results
Alkaline impurities | Passes test |
Acid value | Max. 0.3 |
Peroxide value | Max. 10.0 |
Fatty acid composition | Passes test |
Heavy metals | Max. 10 µg/g |
Sterol composition (as Brassicasterol) | Max. 0.3 % |
Water | Max. 0.1% |
Unsaponifiable matter | Max.1.5% |
Identification | Passes test |
Residual solvents | Passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...