Sulfur dioxide, anhydrous ≥99.98%

Supplier: Pfaltz & Bauer
Danger

S12118-454G
101178-996EA 3893.43 USD
101178-996
Sulfur dioxide, anhydrous ≥99.98%
Sulphur dioxide
Formula: O₂S
MW: 64.06 g/mol
Boiling Pt: –10 °C
Melting Pt: –73 °C
CAS Number: 7446-09-5
UN: 1079
ADR: 2,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR